Merck
Regulation of Gluconeogenesis by Aldo-keto-reductase 1a1b in Zebrafish.
iScience (2020-12-01)
Xiaogang Li, Felix Schmöhl, Haozhe Qi, Katrin Bennewitz, Christoph T Tabler, Gernot Poschet, Rüdiger Hell, Nadine Volk, Tanja Poth, Ingrid Hausser, Jakob Morgenstern, Thomas Fleming, Peter Paul Nawroth, Jens Kroll
ABSTRAKT
Regulation of glucose homeostasis is a fundamental process to maintain blood glucose at a physiological level, and its dysregulation is associated with the development of several metabolic diseases. Here, we report on a zebrafish mutant for Aldo-keto-reductase 1a1b (akr1a1b) as a regulator of gluconeogenesis. Adult akr1a1b -/- mutant zebrafish developed fasting hypoglycemia, which was caused by inhibiting phosphoenolpyruvate carboxykinase (PEPCK) expression as rate-limiting enzyme of gluconeogenesis. Subsequently, glucogenic amino acid glutamate as substrate for gluconeogenesis accumulated in the kidneys, but not in livers, and induced structural and functional pronephros alterations in 48-hpf akr1a1b -/- embryos. Akr1a1b -/- mutants displayed increased nitrosative stress as indicated by increased nitrotyrosine, and increased protein-S-nitrosylation. Inhibition of nitrosative stress using the NO synthase inhibitor L-NAME prevented kidney damage and normalized PEPCK expression in akr1a1b -/- mutants. Thus, the data have identified Akr1a1b as a regulator of gluconeogenesis in zebrafish and thereby controlling glucose homeostasis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, for silylations, LiChropur
Supelco
Alkane standard mixture for performance tests of GC-systems, analytical standard, C10 - C40 (all even), 50 mg/l each
Supelco
Trimethylsulfonium hydroxide solution, ~0.25 M in methanol, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
5-Propyl-2-thiouracil, ≥98%
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
Methoxyamine hydrochloride, 98%