Merck
  • Systematic Characterization of Recurrent Genomic Alterations in Cyclin-Dependent Kinases Reveals Potential Therapeutic Strategies for Cancer Treatment.
Systematic Characterization of Recurrent Genomic Alterations in Cyclin-Dependent Kinases Reveals Potential Therapeutic Strategies for Cancer Treatment.
Cell reports (2020-07-16)
Weiwei Shan, Jiao Yuan, Zhongyi Hu, Junjie Jiang, Yueying Wang, Nicki Loo, Lingling Fan, Zhaoqing Tang, Tianli Zhang, Mu Xu, Yutian Pan, Jiaqi Lu, Meixiao Long, Janos L Tanyi, Kathleen T Montone, Yi Fan, Xiaowen Hu, Youyou Zhang, Lin Zhang
ABSTRAKT
Recurrent copy-number alterations, mutations, and transcript fusions of the genes encoding CDKs/cyclins are characterized in >10,000 tumors. Genomic alterations of CDKs/cyclins are dominantly driven by copy number aberrations. In contrast to cell-cycle-related CDKs/cyclins, which are globally amplified, transcriptional CDKs/cyclins recurrently lose copy numbers across cancers. Although mutations and transcript fusions are relatively rare events, CDK12 exhibits recurrent mutations in multiple cancers. Among the transcriptional CDKs, CDK7 and CDK12 show the most significant copy number loss and mutation, respectively. Their genomic alterations are correlated with increased sensitivities to DNA-damaging drugs. Inhibition of CDK7 preferentially represses the expression of genes in the DNA-damage-repair pathways and impairs the activity of homologous recombination. Low-dose CDK7 inhibitor treatment sensitizes cancer cells to PARP inhibitor-induced DNA damage and cell death. Our analysis provides genomic information for identification and prioritization of drug targets for CDKs and reveals rationales for treatment strategies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H2A.X (Ser139), klon JBW301, clone JBW301, Upstate®, from mouse