Merck
  • Ribose-modified purine nucleosides as ribonucleotide reductase inhibitors. Synthesis, antitumor activity, and molecular modeling of N6-substituted 3'-C-methyladenosine derivatives.
Ribose-modified purine nucleosides as ribonucleotide reductase inhibitors. Synthesis, antitumor activity, and molecular modeling of N6-substituted 3'-C-methyladenosine derivatives.
Journal of medicinal chemistry (2008-07-01)
Loredana Cappellacci, Palmarisa Franchetti, Patrizia Vita, Riccardo Petrelli, Antonio Lavecchia, Hiremagalur N Jayaram, Philipp Saiko, Geraldine Graser, Thomas Szekeres, Mario Grifantini
ABSTRAKT
A series of cycloalkyl, bicycloalkyl, aryl, and heteroaryl N (6)-substituted derivatives of the antitumor agent 3'- C-methyladenosine (3'-Me-Ado), an inhibitor of the alpha Rnr1 subunit of mammalian ribonucleotide reductase (RR), were synthesized. The cytotoxicity of these compounds was evaluated against a panel of human leukemia and carcinoma cell lines and compared to that of some corresponding N (6)-substituted adenosine analogues. N (6)-cycloalkyl-3'- C-methylribonucleosides 2- 7 and N (6)-phenyl analogue 8 were found to inhibit the proliferation of K562 leukemia cells. N (6)-(+/-)- endo-2-norbornyl-3'- C-methyladenosine ( 7) was found to be the most cytotoxic compound, with GI 50 values slightly higher than that of 3'-Me-Ado against K562 and carcinoma cell lines and 2.7 fold higher cytotoxicity against human promyelocytic leukemia HL-60 cells. The SAR study confirms that an unsubstituted N (6)-amino group is essential for optimal cytotoxicity of 3'-Me-Ado against both K562 and carcinoma cell lines. Computational studies, carried out on the eukaryotic alpha subunit (Rnr1) of RR from Saccharomyces cerevisiae were performed to rationalize the observed structure-activity relationships.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Chloro-N6-cyclopentyladenosine, adenosine receptor agonist