Merck
The uptake of tau amyloid fibrils is facilitated by the cellular prion protein and hampers prion propagation in cultured cells.
Journal of neurochemistry (2020-05-13)
Elena De Cecco, Luigi Celauro, Silvia Vanni, Micaela Grandolfo, Edoardo Bistaffa, Fabio Moda, Adriano Aguzzi, Giuseppe Legname
ABSTRAKT
Tauopathies are prevalent, invariably fatal brain diseases for which no cure is available. Tauopathies progressively affect the brain through cell-to-cell transfer of tau protein amyloids, yet the spreading mechanisms remain unknown. Here we show that the cellular prion protein (PrPC ) facilitates the uptake of tau aggregates by cultured cells, possibly by acting as an endocytic receptor. In mouse neuroblastoma cells, pull-down experiments revealed that tau amyloids bind to PrPC . Confocal images of both wild-type and PrPC -knockout N2a cells treated with fluorescently labeled synthetic tau fibrils showed that the internalization was reduced in isogenic cells devoid of the gene encoding PrPC . Pre-treatment of the same cells with antibodies against N-proximal epitopes of PrPC impaired the binding of tau amyloids and decreased their uptake. Surprisingly, exposure of chronically prion-infected cells to tau amyloids reduced the accumulation of aggregated prion protein and this effect lasted for more than 72 hr after amyloid removal. These results point to bidirectional interactions between the two proteins: while PrPC mediates the entrance of tau fibrils in cells, PrPSc buildup is greatly reduced in their presence, possibly because of an impairment in the prion conversion process.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture