Merck
Cinnamoyl- and hydroxycinnamoyl amides of glaucine and their antioxidative and antiviral activities.
Bioorganic & medicinal chemistry (2008-07-02)
Maya Spasova, Stefan Philipov, L Nikolaeva-Glomb, A S Galabov, Ts Milkova
ABSTRAKT
The aporphine alkaloid glaucine has been converted into 3-aminomethylglaucine and its free amino group has been linked to cinnamic, ferulic, sinapic, o-, and p-coumaric acids. The antioxidative potential of the synthesized amides was studied against DPPH(*) test. All of the tested compounds demonstrated higher radical scavenging activity than glaucine and 3-aminomethylglaucine, and lower antioxidative effect than the free hydroxycinnamic acids. The newly synthesized compounds were tested in vitro for antiviral activity against viruses belonging to different taxonomic groups.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland