Merck
First-in-class humanized FSH blocking antibody targets bone and fat.
Proceedings of the National Academy of Sciences of the United States of America (2020-11-01)
Sakshi Gera, Damini Sant, Shozeb Haider, Funda Korkmaz, Tan-Chun Kuo, Mehr Mathew, Helena Perez-Pena, Honglin Xie, Hao Chen, Rogerio Batista, Kejun Ma, Zhen Cheng, Elina Hadelia, Cemre Robinson, Anne Macdonald, Sari Miyashita, Anthony Williams, Gregory Jebian, Hirotaka Miyashita, Anisa Gumerova, Kseniia Ievleva, Pinar Smith, Jiahuan He, Vitaly Ryu, Victoria DeMambro, Matthew A Quinn, Marcia Meseck, Se-Min Kim, T Rajendra Kumar, Jameel Iqbal, Maria I New, Daria Lizneva, Clifford J Rosen, Aaron J Hsueh, Tony Yuen, Mone Zaidi
ABSTRAKT
Blocking the action of FSH genetically or pharmacologically in mice reduces body fat, lowers serum cholesterol, and increases bone mass, making an anti-FSH agent a potential therapeutic for three global epidemics: obesity, osteoporosis, and hypercholesterolemia. Here, we report the generation, structure, and function of a first-in-class, fully humanized, epitope-specific FSH blocking antibody with a K D of 7 nM. Protein thermal shift, molecular dynamics, and fine mapping of the FSH-FSH receptor interface confirm stable binding of the Fab domain to two of five receptor-interacting residues of the FSHβ subunit, which is sufficient to block its interaction with the FSH receptor. In doing so, the humanized antibody profoundly inhibited FSH action in cell-based assays, a prelude to further preclinical and clinical testing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Leukocyte Acid Phosphatase (TRAP) Kit, Select reagents packaged in gelatin capsules
Millipore
MILLIPLEX® Mouse Pituitary Panel, Configurable Mouse Pituitary 7-Plex Panel
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder