Merck
Impaired T cell-mediated hepatitis in peroxisome proliferator activated receptor alpha (PPARα)-deficient mice.
Biological research (2018-02-17)
Ian N Hines, Michael Kremer, Sherri M Moore, Michael D Wheeler
ABSTRAKT
Peroxisome proliferator activated receptor alpha (PPARα), a regulator of enzymes involved in β oxidation, has been reported to influence lymphocyte activation. The purpose of this study was to determine whether PPARα plays a role in T cell-mediated hepatitis induced by Concanavalin A (ConA). Wild type (wt) or PPARα-deficient (PPARα-/-) mice were treated with ConA (15 mg/kg) by intravenous injection 0, 10 or 24 h prior to sacrifice and serum and tissue collection for analysis of tissue injury, cytokine response, T cell activation and characterization. Ten and 24 h following ConA administration, wt mice had significant liver injury as demonstrated by serum transaminase levels, inflammatory cell infiltrate, hepatocyte apoptosis, and expression of several cytokines including interleukin 4 (IL4) and interferon gamma (IFNγ). In contrast, PPARα-/- mice were protected from ConA-induced liver injury with significant reductions in serum enzyme release, greatly reduced inflammatory cell infiltrate, hepatocellular apoptosis, and IFNγ expression, despite having similar levels of hepatic T cell activation and IL4 expression. This resistance to liver injury was correlated with reduced numbers of hepatic natural killer T (NKT) cells and their in vivo responsiveness to alpha-galactosylceramide. Interestingly, adoptive transfer of either wt or PPARα-/- splenocytes reconstituted ConA liver injury and cytokine production in lymphocyte-deficient, severe combined immunodeficient mice implicating PPARα within the liver, possibly through support of IL15 expression and/or suppression of IL12 production and not the lymphocyte as the key regulator of T cell activity and ConA-induced liver injury. Taken together, these data suggest that PPARα within the liver plays an important role in ConA-mediated liver injury through regulation of NKT cell recruitment and/or survival.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides
Roche
Zestaw do wykrywania śmierci komórek in situ, fluoresceina, sufficient for ≤50 tests, suitable for detection