Merck
MechanoBioTester: A Decoupled Multistimulus Cell Culture Device for Studying Complex Microenvironments In Vitro.
ACS biomaterials science & engineering (2020-07-25)
Bryan D James, Nicolas Montoya, Josephine Allen
ABSTRAKT
Increasingly being recognized is the role of the complex microenvironment to regulate cell phenotype; however, the cell culture systems used to study these effects in vitro are lagging. The complex microenvironment is host to a combination of biological interactions, chemical factors, and mechanical stimuli. Many devices have been designed to probe the effects of one mechanical stimulus, but few are capable of systematically interrogating all combinations of mechanical stimuli with independent control. To address this gap, we have developed the MechanoBioTester platform, a decoupled, multi-stimulus cell culture model for studying the cellular response to complex microenvironments in vitro. The system uses an engineered elastomeric chamber with a specially defined region for incorporating different target materials to act as the cell culture substrate. We have tested the system with several target materials including: polydimethylsiloxane elastomer, polyacrylamide gel, poly(1,8-octanediol citrate) elastomer, and type I collagen gel for both 2D and 3D co-culture. Additionally, when the chamber is connected to a flow circuit and our stretching device, stimuli in the form of fluid flow, cyclic stretch, and hydrostatic pressure are able to be imparted with independent control. We validated the device using experimental and computational methods to define a range of capabilities relevant to physiological microenvironments. The MechanoBioTester platform promises to function as a model system for mechanobiology, biomaterial design, and drug discovery applications that focus on probing the impact of a complex microenvironment in an in vitro setting. The protocol described within provides the details characterizing the MechanoBioTester system, the steps for fabricating the MechanoBioTester chamber, and the procedure for operating the MechanoBioTester system to stimulate cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
1,8-Octanediol, 98%