Merck
Laterally Aggregated Polyacrylamide Gels for Immunoprobed Isoelectric Focusing.
Analytical chemistry (2020-01-28)
Shaheen Jeeawoody, Kevin A Yamauchi, Alison Su, Amy E Herr
ABSTRAKT
Immunoprobed isoelectric focusing (IEF) resolves proteins based on differences in isoelectric point (pI) and then identifies protein targets through immunoprobing of IEF-separated proteins that have been immobilized onto a gel scaffold. During the IEF stage, the gel functions as an anti-convective medium and not as a molecular sieving matrix. During the immunoprobing stage, the gel acts as an immobilization scaffold for IEF-focused proteins via photoactive moieties. Here, we characterized the effect of gel pore size on IEF separation and in-gel immunoassay performance. We modulated polyacrylamide (PA) gel pore size via lateral chain aggregation initiated by PEG monomers. During IEF, the 2% PEG highly porous PA gel formulation offered higher resolution (minimum pI difference ∼0.07 ± 0.02) than unmodified 6%T, 3.3%C (benchmark) and 6%T, 8%C (negative control) PA gels. The highly porous gels supported a pH gradient with slope and linearity comparable to benchmark gels. The partition coefficient for antibodies into the highly porous gels (K = 0.35 ± 0.02) was greater than the benchmark (3×) and negative control (1.75×) gels. The highly porous gels also had lower immunoassay background signal than the benchmark (2×) and negative control (3×) gels. Taken together, lateral aggregation creates PA gels that are suitable for both IEF and subsequent in-gel immunoprobing by mitigating immunoprobe exclusion from the gels while facilitating removal of unbound immunoprobe.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%