Merck
TMEM173 Drives Lethal Coagulation in Sepsis.
Cell host & microbe (2020-03-07)
Hui Zhang, Ling Zeng, Min Xie, Jiao Liu, Borong Zhou, Runliu Wu, Lizhi Cao, Guido Kroemer, Haichao Wang, Timothy R Billiar, Herbert J Zeh, Rui Kang, Jianxin Jiang, Yan Yu, Daolin Tang
ABSTRAKT
The discovery of TMEM173/STING-dependent innate immunity has recently provided guidance for the prevention and management of inflammatory disorders. Here, we show that myeloid TMEM173 occupies an essential role in regulating coagulation in bacterial infections through a mechanism independent of type I interferon response. Mechanistically, TMEM173 binding to ITPR1 controls calcium release from the endoplasmic reticulum in macrophages and monocytes. The TMEM173-dependent increase in cytosolic calcium drives Gasdermin D (GSDMD) cleavage and activation, which triggers the release of F3, the key initiator of blood coagulation. Genetic or pharmacological inhibition of the TMEM173-GSDMD-F3 pathway blocks systemic coagulation and improves animal survival in three models of sepsis (cecal ligation and puncture or bacteremia with Escherichia coli or Streptococcus pneumoniae infection). The upregulation of the TMEM173 pathway correlates with the severity of disseminated intravascular coagulation and mortality in patients with sepsis. Thus, TMEM173 is a key regulator of blood clotting during lethal bacterial infections.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Millipore
Tauroursodeoxycholic Acid, Sodium Salt, Tauroursodeoxycholic is a detergent useful for the solubilization of lipids and membrane-bound proteins.
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
2-APB, A cell-permeable modulator of Ins(1,4,5)P3-induced Ca2+ release.
Sigma-Aldrich
U-73122, U-73122, CAS 112648-68-7, inhibits agonist-induced phospholipase C activation (IC50 = 1-2.1 µM) in human platelets and neutrophils.