Merck
Repulsive Guidance Molecule A Suppresses Adult Neurogenesis.
Stem cell reports (2020-04-04)
Toke Jost Isaksen, Yuki Fujita, Toshihide Yamashita
ABSTRAKT
Repulsive guidance molecule A (RGMa) is a glycosylphosphatidylinositol-anchored glycoprotein that exhibits repulsive neurite guidance and regulates neuronal differentiation and survival during brain development. However, the function of RGMa in the adult brain is unknown. Here, we show that RGMa is expressed in the adult hippocampus and provide evidence that RGMa signaling suppresses adult neurogenesis. Knockdown of RGMa in the dentate gyrus increased the number of surviving newborn neurons; however, these cells failed to properly migrate into the granular cell layer. In vitro, RGMa stimulation of adult neural stem cells suppressed neurite outgrowth of newborn neurons, which could be prevented by knockdown of the multifunctional receptor neogenin, as well as pharmacological inhibition of the downstream target Rho-associated protein kinase. These findings present a function for RGMa in the adult brain and add to the intricate molecular network that regulates adult brain plasticity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 10 ×, sterile-filtered, BioReagent, suitable for cell culture, 5.0 g porcine trypsin and 2 g EDTA, 4Na per liter of 0.9% sodium chloride
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder