Merck
Antinociceptive properties of caffeic acid derivatives in mice.
European journal of medicinal chemistry (2009-07-25)
Fátima de Campos Buzzi, Caroline Liandra Franzoi, Graziele Antonini, Mauricio Fracasso, Valdir Cechinel Filho, Rosendo Augusto Yunes, Rivaldo Niero
ABSTRAKT
Ten ester derivatives from caffeic acid were synthesized, and their antinociceptive properties are evaluated in mice. The most active compound, dodecyl ester derivative, exhibited potent and dose-related activity against the writhing test, with a calculated ID(50) value of 15.1 (11.9-19.1)micromol/kg and MI of 78.8% being several times more active than reference drugs. It was also effective in other experimental models, such as formalin, capsaicin and glutamate-induced pain tests, but was inactive in the hot-plate test. Although the mechanism of action has still not been elucidated, these results appear to support its therapeutic potential against painful diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder