Merck
  • Influence of Pasteurization and Storage on Dynamic In Vitro Gastric Digestion of Milk Proteins: Quantitative Insights Based on Peptidomics.
Influence of Pasteurization and Storage on Dynamic In Vitro Gastric Digestion of Milk Proteins: Quantitative Insights Based on Peptidomics.
Foods (Basel, Switzerland) (2020-07-30)
Xing Li, Yuxiang Gu, Shudong He, Olayemi Eyituoyo Dudu, Qiming Li, Haiyan Liu, Ying Ma
ABSTRAKT
It is important to evaluate the nutritional quality of milk during the shelf-life, especially during home storage, from a consumer viewpoint. In this study, we investigated the impact of pasteurization (85 °C/15 s) and subsequent storage (at 4 °C for 7 days) on the coagulation behavior of milk and protein digestibility in a dynamic in vitro gastric digestion test. A high level of hydration in curd formed in pasteurized milk upon 7-day cold storage compared to raw and pasteurized milk, indicating fast pepsin diffusion in the interior of curds, increasing the hydrolysis rate. The digesta collected at various time points throughout the gastric digestion were studied using o-phthaldialdehyde (OPA), sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE), liquid chromatography tandem mass spectrometry (LC-MS/MS), and amino acid analysis. These results showed that milk proteins were hydrolyzed quickly upon a long period of cold storage. Additionally, qualitative and quantitative results obtained using LC-MS/MS exhibited significant differences between samples, especially in pasteurized milk upon cold storage. Processing and storage played a decisive role in bioactive peptide generation. Such knowledge could provide insights into and directions for the storage of pasteurized milk for further clinical studies on protein bioavailability and the generation of bioactive peptides for desired health outcomes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid