Merck
  • Occurrence of efflux mechanism and cephalosporinase variant in a population of Enterobacter aerogenes and Klebsiella pneumoniae isolates producing extended-spectrum beta-lactamases.
Occurrence of efflux mechanism and cephalosporinase variant in a population of Enterobacter aerogenes and Klebsiella pneumoniae isolates producing extended-spectrum beta-lactamases.
Antimicrobial agents and chemotherapy (2009-01-24)
Que-Tien Tran, Myrielle Dupont, Jean-Philippe Lavigne, Jacqueline Chevalier, Jean-Marie Pagès, Albert Sotto, Anne Davin-Regli
ABSTRAKT
We investigated the occurrence of multidrug resistance in 44 Enterobacter aerogenes and Klebsiella pneumoniae clinical isolates. Efflux was involved in resistance in E. aerogenes isolates more frequently than in K. pneumoniae isolates (100 versus 38% of isolates) and was associated with the expression of phenylalanine arginine beta-naphthylamide-susceptible active efflux. AcrA-TolC overproduction in E. aerogenes isolates was noted. An analysis of four E. aerogenes isolates for which cefepime MICs were high revealed no modification in porin expression but a new specific mutation in the AmpC beta-lactamase.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)