Merck
ACHN-490, a neoglycoside with potent in vitro activity against multidrug-resistant Klebsiella pneumoniae isolates.
Antimicrobial agents and chemotherapy (2009-09-23)
Andrea Endimiani, Kristine M Hujer, Andrea M Hujer, Eliana S Armstrong, Yuvraj Choudhary, James B Aggen, Robert A Bonomo
ABSTRAKT
The in vitro activity of ACHN-490, a novel aminoglycoside ("neoglycoside"), was evaluated against 102 multidrug-resistant (MDR) Klebsiella pneumoniae strains, including a subset of 25 strains producing the KPC carbapenemase. MIC50 values for gentamicin, tobramycin, and amikacin were 8 microg/ml, 32 microg/ml, and 2 microg/ml, respectively; MIC90 values for the same antimicrobials were > or = 64 microg/ml, > or = 64 microg/ml, and 32 microg/ml, respectively. ACHN-490 showed an MIC50 of 0.5 microg/ml and an MIC90 of 1 microg/ml, which are significantly lower than those of comparator aminoglycosides. ACHN-490 represents a promising aminoglycoside for the treatment of MDR K. pneumoniae isolates, including those producing KPC beta-lactamase.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)