Merck
  • System Prediction and Validation of TCM for Chronic Myeloid Leukemia Treatment from the Perspective of Low-Toxicity Chemotherapy: A Stilbene α-Viniferin Has a Proapoptotic Effect on K562 Cells via the Mitochondrial Pathway.
System Prediction and Validation of TCM for Chronic Myeloid Leukemia Treatment from the Perspective of Low-Toxicity Chemotherapy: A Stilbene α-Viniferin Has a Proapoptotic Effect on K562 Cells via the Mitochondrial Pathway.
Evidence-based complementary and alternative medicine : eCAM (2020-02-28)
Bing-Yang Chai, Fu-Kai Gong, Ze-Hui Chen, Zhao-Xue Li, Bo Zhang
ABSTRAKT
In traditional Chinese medicine (TCM), chronic myeloid leukemia (CML) has been attributed to "poisoned bone marrow," which is viewed as a loss of Qi or blood, a deficiency in Yin or Yang that causes a complex imbalance between cell growth and death. Malignant myeloid progenitor cells display excessive growth that is difficult to control without toxicity. More than 60 herbs in TCM have shown efficacy against CML. However, the key molecules and mechanisms involved in the holistic-level characterization, as well as the effective target associations, are still unknown. The present study employed a computational approach with filtering potential compounds via admetSAR, systems biology-based functional data prediction, and biochemical and molecular biological validation. We generated 118 bioactive compounds from 11 herbs within four dialectical therapy groups that are most commonly used to treat CML and predicted 141 potential targets. The stilbene resveratrol and its derivatives were found to be highly related to these targets. Among them, α-viniferin was predicted to target Bcl-2, caspase-3, 8, and 9, MAPK14, CDK2, HSP90AA1, and others, reflecting CML therapeutic strategies. In vitro, experimental data showed a nonnecrotic growth limitation of K562 cells caused by α-viniferin was predicted to target Bcl-2, caspase-3, 8, and 9, MAPK14, CDK2, HSP90AA1, and others, reflecting CML therapeutic strategies. μg·mL-1 at 24 h. Finally, we validated the chemotherapeutic effect of α-viniferin was predicted to target Bcl-2, caspase-3, 8, and 9, MAPK14, CDK2, HSP90AA1, and others, reflecting CML therapeutic strategies. Our work sheds light on the mechanism of the efficacy of the stilbene α-viniferin in TCM for the prevention of CML. This work also predicts and validates targets in the mitochondrial signaling pathway, providing a novel strategy for CML treatment.α-viniferin was predicted to target Bcl-2, caspase-3, 8, and 9, MAPK14, CDK2, HSP90AA1, and others, reflecting CML therapeutic strategies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)