Merck
Glucose uptake enhancing activity of puerarin and the role of C-glucoside suggested from activity of related compounds.
Bioorganic & medicinal chemistry letters (2010-07-08)
Eisuke Kato, Jun Kawabata
ABSTRAKT
Chemical treatment of diabetes mellitus is widely studied and controlling of blood glucose level is the main course of therapy. In type 2 diabetes mellitus, insulin resistance is the major problem. An isoflavone C-glucoside, puerarin (1), is known to enhance glucose uptake into the insulin sensitive cell and is thought to be a candidate for treatment of diabetes mellitus. We synthesized 1 and several derivatives to apply for the structure-activity relationship study. The result against 3T3-L1 adipocyte indicated that the C-glucoside part of 1 is unconcerned in its activity when tested in vitro and the main structure responsible for its activity was the isoflavone moiety.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Supelco
Puerarin, analytical standard