Merck
  • Synthesis and antimicrobial activity of novel fluorine containing 4-(substituted-2-hydroxybenzoyl)-1H-pyrazoles and pyrazolyl benzo[d]oxazoles.
Synthesis and antimicrobial activity of novel fluorine containing 4-(substituted-2-hydroxybenzoyl)-1H-pyrazoles and pyrazolyl benzo[d]oxazoles.
Bioorganic & medicinal chemistry letters (2010-08-21)
Amol V Gadakh, Chetan Pandit, Sahebrao S Rindhe, Bhausaheb K Karale
ABSTRAKT
A series of fluorine containing 4-(substituted-2-hydroxybenzoyl) pyrazoles and pyrazolyl benzo[d]oxazoles were synthesized and evaluated for their antibacterial activity against Staphylococcus aureus, Escherichia coli, Pseudomonas aeruginosa and Bacillus subtilis and antifungal activity against Candida albicans. The antibacterial activities were expressed as the minimum inhibitory concentration (MIC50) in microg/ml. The compounds 1-(3,4-difluorophenyl)-4-(5-fluoro-2-hydroxybenzoyl)-1H-pyrazole (4b), oxime derivatives such as 1-(3,4-difluorophenyl)-1H-pyrazol-4-yl)(2-hydroxy-4-methylphenyl)methanone oxime (5b) and (5-chloro-2-hydroxyphenyl)(1-(3,4-difluorophenyl)-1H-pyrazol-4-yl)methanone oxime (5e) exhibited promising activities against tested bacterial strains. Except compound 1-(3,4-difluorophenyl)-4-(2-hydroxybenzoyl)-1H-pyrazole (4d), none of the other compounds showed promising antifungal activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard