Merck
Hydrogen sulfide, oxygen, and calcium regulation in developing human airway smooth muscle.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-08-11)
Colleen M Bartman, Marta Schiliro, Martin Helan, Y S Prakash, David Linden, Christina Pabelick
ABSTRAKT
Preterm infants can develop airway hyperreactivity and impaired bronchodilation following supplemental O2 (hyperoxia) in early life, making it important to understand mechanisms of hyperoxia effects. Endogenous hydrogen sulfide (H2 S) has anti-inflammatory and vasodilatory effects with oxidative stress. There is little understanding of H2 S signaling in developing airways. We hypothesized that the endogenous H2 S system is detrimentally influenced by O2 and conversely H2 S signaling pathways can be leveraged to attenuate deleterious effects of O2 . Using human fetal airway smooth muscle (fASM) cells, we investigated baseline expression of endogenous H2 S machinery, and effects of exogenous H2 S donors NaHS and GYY4137 in the context of moderate hyperoxia, with intracellular calcium regulation as a readout of contractility. Biochemical pathways for endogenous H2 S generation and catabolism are present in fASM, and are differentially sensitive to O2 toward overall reduction in H2 S levels. H2 S donors have downstream effects of reducing [Ca2+ ]i responses to bronchoconstrictor agonist via blunted plasma membrane Ca2+ influx: effects blocked by O2 . However, such detrimental O2 effects are targetable by exogenous H2 S donors such as NaHS and GYY4137. These data provide novel information regarding the potential for H2 S to act as a bronchodilator in developing airways in the context of oxygen exposure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydrosulfide hydrate
Sigma-Aldrich
Xestospongin C, film
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Glybenclamide, ≥99% (HPLC)
Sigma-Aldrich
Antimycin A from Streptomyces sp.