Merck
Synthesis of potent, substituted carbazoles as selective androgen receptor modulators (SARMs).
Bioorganic & medicinal chemistry letters (2010-11-03)
Chris P Miller, Pushpal Bhaket, Nagarajan Muthukaman, C Richard Lyttle, Maysoun Shomali, Kyla Gallacher, Connie Slocum, Gary Hattersley
ABSTRAKT
The synthesis and in vitro binding affinity for a novel series of potent androgen receptor modulators is described. One of the more potent compounds (17, RAD35010) was further characterized in vivo where it restored levator ani weight in castrated male rats to near sham level while having no significant effect on prostate weight.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
Testosterone, VETRANAL®, analytical standard