Merck
Carboxymethylated-kappa-casein: a convenient tool for the identification of polyphenolic inhibitors of amyloid fibril formation.
Bioorganic & medicinal chemistry (2009-11-26)
John A Carver, Peter J Duggan, Heath Ecroyd, Yanqin Liu, Adam G Meyer, C Elisabet Tranberg
ABSTRAKT
Reduced and carboxymethylated-kappa-casein (RCM-kappa-CN) is a milk-derived amyloidogenic protein that readily undergoes nucleation-dependent aggregation and amyloid fibril formation via a similar pathway to disease-specific amyloidogenic peptides like amyloid beta (Abeta), which is associated with Alzheimer's disease. In this study, a series of flavonoids, many known to be inhibitors of Abeta fibril formation, were screened for their ability to inhibit RCM-kappa-CN fibrilisation, and the results were compared with literature data on Abeta inhibition. Flavonoids that had a high degree of hydroxylation and molecular planarity gave good inhibition of RCM-kappa-CN fibril formation. IC(50) values were between 10- and 200-fold higher with RCM-kappa-CN than literature results for Abeta fibril inhibition, however, with few exceptions, they showed a similar trend in potency. The convenience and reproducibility of the RCM-kappa-CN assay make it an economic alternative first screen for Abeta inhibitory activity, especially for use with large compound libraries.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)