Merck
  • Exposure of Rats to Multiple Oral Doses of Dichloroacetate Results in Upregulation of Hepatic Glutathione Transferases and NAD(P)H Dehydrogenase [Quinone] 1.
Exposure of Rats to Multiple Oral Doses of Dichloroacetate Results in Upregulation of Hepatic Glutathione Transferases and NAD(P)H Dehydrogenase [Quinone] 1.
Drug metabolism and disposition: the biological fate of chemicals (2020-09-03)
Edwin J Squirewell, Ricky Mareus, Lloyd P Horne, Peter W Stacpoole, Margaret O James
ABSTRAKT
Dichloroacetate (DCA) is an investigational drug that is used in the treatment of various congenital and acquired disorders of energy metabolism. Although DCA is generally well tolerated, some patients experience peripheral neuropathy, a side effect more common in adults than children. Repetitive DCA dosing causes downregulation of its metabolizing enzyme, glutathione transferase zeta 1 (GSTZ1), which is also critical in the detoxification of maleylacetoacetate and maleylacetone. GSTZ1 (-/-) knockout mice show upregulation of glutathione transferases (GSTs) and antioxidant enzymes as well as an increase in the ratio of oxidized glutathione (GSSG) to reduced glutathione (GSH), suggesting GSTZ1 deficiency causes oxidative stress. We hypothesized that DCA-mediated depletion of GSTZ1 causes oxidative stress and used the rat to examine induction of GSTs and antioxidant enzymes after repeated DCA exposure. We determined the expression of alpha, mu, pi, and omega class GSTs, NAD(P)H dehydrogenase [quinone] 1 (NQO1), gamma-glutamylcysteine ligase complex (GCLC), and glutathione synthetase (GSS). GSH and GSSG levels were measured by liquid chromatography-tandem mass spectrometry. Enzyme activity was measured in hepatic cytosol using 1-chloro-2,4-dinitrobenzene, 1,2-dichloro-4-nitrobenzene, and 2,6-dichloroindophenol as substrates. In comparison with acetate-treated controls, DCA dosing increased the relative expression of GSTA1/A2 irrespective of rodent age, whereas only adults displayed higher levels of GSTM1 and GSTO1. NQO1 expression and activity were higher in juveniles after DCA dosing. GSH concentrations were increased by DCA in adults, but the GSH:GSSG ratio was not changed. Levels of GCLC and GSS were higher and lower, respectively, in adults treated with DCA. We conclude that DCA-mediated depletion of GSTZ1 causes oxidative stress and promotes the induction of antioxidant enzymes that may vary between age groups. SIGNIFICANCE STATEMENT: Treatment with the investigational drug, dichloroacetate (DCA), results in loss of glutathione transferase zeta 1 (GSTZ1) and subsequent increases in body burden of the electrophilic tyrosine metabolites, maleylacetoacetate and maleylacetone. Loss of GSTZ1 in genetically modified mice is associated with induction of glutathione transferases and alteration of the ratio of oxidized to reduced glutathione. Therefore, we determined whether pharmacological depletion of GSTZ1 through repeat administration of DCA produced similar changes in the liver, which could affect responses to other drugs and toxicants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Glu-Cys, ≥80% (HPLC)