Merck
Macrolide and clindamycin resistance in Streptococcus milleri group isolates from the airways of cystic fibrosis patients.
Antimicrobial agents and chemotherapy (2010-04-21)
Margot E Grinwis, Christopher D Sibley, Michael D Parkins, Christina S Eshaghurshan, Harvey R Rabin, Michael G Surette
ABSTRAKT
Organisms belonging to the Streptococcus milleri group (SMG) are known for their role in pyogenic infections but have recently been implicated as etiological agents of pulmonary exacerbation in adult patients with cystic fibrosis (CF). The prolonged exposure of CF patients to antibiotics prompted us to investigate the susceptibility profiles of 118 SMG isolates from the airways of CF patients to 12 antibiotics compared to 43 SMG isolates from patients with invasive infections. We found that approximately 60% of all isolates failed to grow using the standard medium for disc diffusion, Mueller-Hinton blood agar (MHBA), so we explored the usefulness of brain heart infusion (BHI) agar for susceptibility testing. Zone-of-inhibition comparisons between BHI and MHBA showed strong correlations for six antibiotics, and interpretations were similar for both medium types. For ceftriaxone and cefepime, both groups of isolates were highly susceptible. Tetracycline resistance levels were comparable between the two groups (22% in CF isolates and 17.4% in invasive isolates). However, more than half of the CF isolates were not susceptible to azithromycin, erythromycin, and clindamycin, compared to 11%, 13%, and 6.5% of invasive isolates, respectively. There were 5-fold and 8-fold increased risks of azithromycin and clindamycin resistance, respectively, for the isolates from the airways of CF patients relative to the invasive isolates. Macrolide resistance was strongly linked to chronic azithromycin therapy in CF patients. This study shows that BHI agar is a suitable alternative for antimicrobial susceptibility testing for the SMG and that SMG isolates from the airways of CF patients are more resistant to macrolides and clindamycin than strains isolated from patients with invasive infections.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg