Merck
Cyclic AMP efflux through MRP4 regulates actin dynamics signalling pathway and sperm motility in bovines.
Scientific reports (2020-09-26)
Nicolás Chiarante, Carlos A I Alonso, Jessica Plaza, Raquel Lottero-Leconte, Camila Arroyo-Salvo, Agustín Yaneff, Claudia E Osycka-Salut, Carlos Davio, Marcelo Miragaya, Silvina Perez-Martinez
ABSTRAKT
Previously we demonstrated that multidrug resistance-associated protein 4 transporter (MRP4) mediates cAMP efflux in bovine spermatozoa and that extracellular cAMP (ecAMP) triggers events associated to capacitation. Here, we deepen the study of the role of MRP4 in bovine sperm function by using MK571, an MRP4 inhibitor. The incubation of spermatozoa with MK571 during 45 min inhibited capacitation-associated events. MRP4 was localized in post-acrosomal region and mid-piece at 15 min capacitation, while at 45 min it was mainly located in the acrosome. After 15 min, MK571 decreased total sperm motility (TM), progressive motility (PM) and several kinematic parameters. The addition of ecAMP rescued MK571 effect and ecAMP alone increased the percentage of motile sperm and kinematics parameters. Since actin cytoskeleton plays essential roles in the regulation of sperm motility, we investigated if MRP4 activity might affect actin polymerization. After 15 min capacitation, an increase in F-actin was observed, which was inhibited by MK571. This effect was reverted by the addition of ecAMP. Furthermore, ecAMP alone increased F-actin levels while no F-actin was detected with ecAMP in the presence of PKA inhibitors. Our results support the importance of cAMP efflux through MRP4 in sperm capacitation and suggest its involvement in the regulation of actin polymerization and motility.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Selektywny suplement chlortetracykliny, suitable for microbiology
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder