Merck
Lipophilicity of acidic compounds: impact of ion pair partitioning on drug design.
Bioorganic & medicinal chemistry letters (2011-05-24)
Mark C Wenlock, Patrick Barton, Tim Luker
ABSTRAKT
In drug discovery projects the ability to show a relationship between a compound's molecular structure and its pharmacokinetic, in vivo efficacy, or toxicity profile is paramount for the design of better analogues. To aid this understanding the measurement of distribution coefficients at some physiologically relevant pH, for example, log D(7.4), is common practice as they are used as a key descriptor in mathematical models for predicting various biological parameters. Evidence is presented that under typical experimental conditions ion pair partitioning can contribute greatly to log D(7.4) results for acidic compounds; if this is ignored it may compromise data analysis within drug discovery projects where the modulation of lipophilicity is a primary design strategy. The work herein focuses on acidic compounds and reflects the experience of AstraZeneca R&D Charnwood (AZ) where ion pair partitioning contributions can be minimized by the routine measurement of log D(5.5) data. The magnitude of ion pair partitioning contributions to the log D(7.4) measurements of 24 acidic drugs are investigated, and the risks to drug discovery projects that ignore such contributions are discussed. The superiority of measured lipophilicity data over calculated data for a set of AZ proprietary acidic compounds is also presented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chlorambucil
Supelco
Sulfamethizole, analytical standard, ≥99% (HPLC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Glipizide, solid
Supelco
Sulfamethizole, VETRANAL®, analytical standard
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Furosemide