Merck
Beta-adrenoceptor antagonist activities and binding affinities of timolol enantiomers in rat atria.
The Journal of pharmacy and pharmacology (1989-09-01)
S Karhuvaara, T Kaila, R Huupponen
ABSTRAKT
S-Timolol is an effective anti-glaucoma drug, but has potentially hazardous side effects. Recently, R-timolol, also, has been reported to be effective in lowering elevated intraocular pressure. In the present study, the beta-adrenoceptor antagonist activities and binding of R- and S-enantiomers of timolol have been examined on rat atrial preparations. The beta-antagonistic activities were investigated using spontaneously beating rat heart atria. Both timolol enantiomers inhibited (-)-isoprenaline-induced chronotropic action competitively. S-Timolol was about 54 times more potent than R-timolol. The apparent binding affinities of timolol enantiomers to beta 1- and beta 2-adrenoceptors were determined by a radioligand binding assay using (-)-[125I]iodocyanopindolol (ICYP) as a marker and CGP 20712 A as a beta 1- and ICI 118,551 as a beta 2-adrenoceptor antagonist. Both enantiomers of timolol inhibited ICYP binding in nanomolar concentrations with Hill coefficients near unity. Neither enantiomer showed selectivity between beta 1- and beta 2-adrenoceptors, but R-timolol was approximately 30 times less active than S-timolol. It is concluded that R-timolol is a relatively potent non-selective beta-adrenoceptor blocking agent, but may possibly exert a more localized beta-adrenoceptor action in the eye than S-timolol, thus improving the safety of ocular timolol therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Timolol maleate salt, ≥98% (TLC), powder