Merck
  • Synthesis of novel N-(2-hydroxy-2-p-tolylethyl)-amide and N-(2-oxo-2-p-tolylethyl)-amide derivatives and their antidyslipidemic and antioxidant activity.
Synthesis of novel N-(2-hydroxy-2-p-tolylethyl)-amide and N-(2-oxo-2-p-tolylethyl)-amide derivatives and their antidyslipidemic and antioxidant activity.
Bioorganic & medicinal chemistry letters (2011-09-21)
T Narender, K Rajendar, S Sarkar, V K Singh, Upma Chaturvedi, A K Khanna, G Bhatia
ABSTRAKT
In continuation of our drug discovery program on metabolic diseases, we identified an alkaloidal amide, that is, Aegeline (V) from the plant Aegle marmelos leaves as a dual acting agent (antihyperlipidemic and antihyperglycemic). We therefore synthesized a series of alkaloidal amides [N-(2-hydroxy-2-p-tolylethyl)-amides and N-(2-oxo-2-p-tolylethyl)-amide derivatives] related to Aegeline and screened for their in vivo antihyperlipidemic activity in Triton induced hyperlipidemia model. The synthetic compounds 4, 17 and 20 showed equipotent activity to the natural product, that is, Aegeline (V). These compounds also showed strong antioxidant activity, which support their antihyperlipidemic activity. Compound 12 showed better antihyperlipidemic and antioxidant profile than the natural product V.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g