Merck
A drug cocktail for protecting against ischemia-reperfusion injury.
Frontiers in bioscience (Landmark edition) (2019-10-06)
Anna Krzywonos-Zawadzka, Mieczyslaw Wozniak, Grzegorz Sawicki, Iwona Bil-Lula
ABSTRAKT
Ischemia followed by reperfusion (I/R) of cardiomyocytes causes release of a large amount of inducible nitric oxide (NO) synthase (iNOS) followed by an increase of asymmetric dimethylarginine (ADMA). ADMA disrupts NO signaling by switching of the NOS activity from NO to the production of reactive oxygen species (ROS). Previously, we have shown that pretreatment of the hearts by co-administration of sub-threshold concentrations of doxycycline, a matrix metalloproteinase (MMPs) inhibitor, ML-7 an inhibitor of myosin light-chain kinase (MLCK) and L-NAME a non-selective NOS inhibitor protects the heart against I/R injury. In this study, we replaced the L-NAME with 1400W (selective inhibitor of iNOS) in the drug cocktail that was Langendorff-perfused into the hearts of Wistar rats before (prevention) or after (treatment) the induction of I/R. This pre-treatment resulted in full protection of contractility, decreased production of iNOS and ADMA and normalized the bioavailability of NO in the I/R hearts. Thus, the formulated drug cocktail protects the heart from I/R injury.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
1400W dihydrochloride, >98%, solid