Merck
Protective effect of ε-viniferin on β-amyloid peptide aggregation investigated by electrospray ionization mass spectrometry.
Bioorganic & medicinal chemistry (2011-04-29)
Tristan Richard, Pascal Poupard, Merian Nassra, Yorgos Papastamoulis, Marie-Laure Iglésias, Stéphanie Krisa, Pierre Waffo-Teguo, Jean-Michel Mérillon, Jean-Pierre Monti
ABSTRAKT
Abnormal β-amyloid peptide accumulation and aggregation is considered to be responsible for the formation and cerebral deposition of senile plaques in the brains of patients with Alzheimer's disease (AD). Inhibition of the formation of β-amyloid (Aβ) fibrils would be an attractive therapeutic target for the treatment of AD. Resveratrol and its derivatives exhibit a broad range of pharmacological properties such as protection against cardiovascular diseases and cancers, as well as promoting antiaging effects. We reported previously that ε-viniferin glucoside (VG), a resveratrol-derived dimer, strongly inhibits Aβ (25-35) fibril formation in vitro. In this study, we investigated the effects of VG on the aggregation of the full-length peptides (Aβ (1-40) and Aβ (1-42)) and on the β-amyloid-induced toxicity in PC12 cells. VG inhibited Aβ cytotoxicity and the non-covalent complex between VG and Aβ was observed by electrospray ionization mass spectrometry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)