Merck
Human gingiva-derived mesenchymal stem cells are therapeutic in lupus nephritis through targeting of CD39-CD73 signaling pathway.
Journal of autoimmunity (2020-06-23)
Junlong Dang, Zhenjian Xu, Anping Xu, Yan Liu, Qingling Fu, Julie Wang, Feng Huang, Yuejuan Zheng, Guangying Qi, Boqing Sun, Joseph A Bellanti, Umadevi Kandalam, Hany A Emam, Wael Jarjour, Song Guo Zheng
ABSTRAKT
Cell specific and cytokine targeted therapeutics have underperformed in systemic lupus erythematosus (SLE). Mesenchymal stem cells (MSCs) have emerged as a novel therapy to address the dysregulation in autoimmune diseases but also have limitations. Human gingiva derived MSCs (GMSCs) are superior in regulating immune responses. Here, we demonstrate that the adoptive transfer of GMSCs homes to and maintains in the kidney and has a robust therapeutic effect in a spontaneous lupus nephritis model. Specifically, GMSCs limits the development of autoantibodies as well as proteinuria, decreases the frequency of plasma cells and lupus nephritis histopathological scores by directly suppressing B cells activation, proliferation and differentiation. The blockage of CD39-CD73 pathway dramatically abrogates the suppressive capacities of GMSCs in vitro and in vivo and highlights the significance of this signaling pathway in SLE. Collectively, manipulation of GMSCs provides a promising strategy for the treatment of patients with SLE and other autoimmune diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alloxazine, 96%
Sigma-Aldrich
NS-398, ≥98% (HPLC), solid
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications