Merck
  • CB1 receptor-dependent desensitisation of TRPV1 channels contributes to the analgesic effect of dipyrone in sensitised primary sensory neurons.
CB1 receptor-dependent desensitisation of TRPV1 channels contributes to the analgesic effect of dipyrone in sensitised primary sensory neurons.
British journal of pharmacology (2020-06-21)
Gilson Goncalves Dos Santos, Ruihui Li, Melissa Pui Een Ng, Julia Borges Paes Lemes, Willians Fernando Vieira, Istvan Nagy, Cláudia Herrera Tambeli, Carlos Amilcar Parada
ABSTRAKT
While dipyrone is a widely used analgesic, its mechanism of action is not completely understood. Recently, we have reported that the dipyrone metabolite 4-aminoantipyrine (4-AA) reduces PGE2 -induced pain-related behaviour through cannabinoid CB1 receptors. Here, we ascertained, in naive and PGE2 -induced "inflamed" conditions, both in vivo and in vitro, the molecular mechanisms involved in the 4-AA-induced analgesic effects. The effect of local administration of 4-AA (160 μg per paw) on capsaicin (0.12 μg per paw) injection-induced pain-related behaviour and 4-AA's effect on 500-nM capsaicin-induced changes in intracellular calcium concentration ([Ca2+ ]i ) in cultured primary sensory neurons were assessed in vivo and in vitro, respectively. 4-AA reduced capsaicin-induced nociceptive behaviour in naive and inflamed conditions through CB1 receptors. 4-AA (100 μM) reduced capsaicin-induced increase in [Ca2+ ]i in a CB1 receptor-dependent manner, when PGE2 was not present. Following PGE2 application, 4-AA (1-50 μM) increased the [Ca2+ ]i . Although 4-AA activated both TRPV1 and TRPA1 channels, increased [Ca2+ ]i was mediated through TRPV1 channels. Activation of TRPV1 channels resulted in their desensitisation. Blocking CB1 receptors reduced both the excitatory and desensitising effects of 4-AA. CB1 receptor-mediated inhibition of TRPV1 channels and TRPV1-mediated Ca2+ -influx- and CB1 receptor-dependent desensitisation of TRPV1 channels contribute to the anti-nociceptive effect of 4-AA in naive and inflamed conditions respectively. Agonists active at both CB1 receptors and TRPV1 channels might be useful as analgesics, particularly in inflammatory conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Capsazepine, ≥98% (HPLC), solid
Sigma-Aldrich
AM251, >98% (HPLC), solid