Merck
Removing nucleic acids from nucleoid-associated proteins purified by affinity column.
Journal of biological methods (2016-01-30)
Audrey Vingadassalon, Philippe Bouloc, Sylvie Rimsky
ABSTRAKT
In bacteria, DNA is tightly compacted in a supercoiled organization, which is mediated in part by nucleoid-associated proteins (NAPs). NAPs are well characterized for their ability to bind nucleic acids and for their involvement in gene regulation. A method commonly used to study protein-nucleic acid interactions involves immunoprecipitation of the protein of interest which is subsequently incubated with nucleic acids. A common cause of artifact is due to nucleic acids that remains bound to the protein of interest during the whole purification process. We developed an optimized method for the purification of tagged NAPs on affinity columns. The combination of three known methods allows removal of most of the nucleic acids bound to proteins during the purification process. This protocol is designed to improve the quality and specificity of results of in vitro experiments involving nucleic acid binding tests on purified NAPs. It can be used for in vitro studies of other RNA/DNA binding proteins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for use with bacterial cell extracts, lyophilized powder
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution