Merck
Synthesis of an Hsp70 inhibitor and its assessment of lysosomal membrane permeabilization.
STAR protocols (2021-03-06)
Sang-Hyun Park, Sookil Park, Injae Shin
ABSTRAKT
Hsp70 inhibitors have great potential as chemical probes and anticancer agents. Thus, it is important to elucidate their modes of action on cancer cell death. This protocol describes a step-by-step process for the synthesis of apoptozole as an inhibitor of Hsp70, analysis of internalization of apoptozole into lysosomes, and assessment of lysosomal membrane permeabilization induced by apoptozole. The current protocol can be used for detailed mechanistic studies of Hsp70 inhibitors and further substances targeting lysosomal proteins on cancer cell death. For complete information on the use and execution of this protocol, please refer to Park et al. (2018).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Roche
cOmplete, Mini koktajl inhibitorów proteaz, Tablets provided in a glass vial
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)