Merck
Accuracy of serological testing for SARS-CoV-2 antibodies: First results of a large mixed-method evaluation study.
Allergy (2020-10-01)
Daniel Brigger, Michael P Horn, Luke F Pennington, Abigail E Powell, Denise Siegrist, Benjamin Weber, Olivier Engler, Vanja Piezzi, Lauro Damonti, Patricia Iseli, Christoph Hauser, Tanja K Froehlich, Peter M Villiger, Martin F Bachmann, Stephen L Leib, Pascal Bittel, Martin Fiedler, Carlo R Largiadèr, Jonas Marschall, Hanspeter Stalder, Peter S Kim, Theodore S Jardetzky, Alexander Eggel, Michael Nagler
ABSTRAKT
Serological immunoassays that can identify protective immunity against SARS-CoV-2 are needed to adapt quarantine measures, assess vaccination responses, and evaluate donor plasma. To date, however, the utility of such immunoassays remains unclear. In a mixed-design evaluation study, we compared the diagnostic accuracy of serological immunoassays that are based on various SARS-CoV-2 proteins and assessed the neutralizing activity of antibodies in patient sera. Consecutive patients admitted with confirmed SARS-CoV-2 infection were prospectively followed alongside medical staff and biobank samples from winter 2018/2019. An in-house enzyme-linked immunosorbent assay utilizing recombinant receptor-binding domain (RBD) of the SARS-CoV-2 spike protein was developed and compared to three commercially available enzyme-linked immunosorbent assays (ELISAs) targeting the nucleoprotein (N), the S1 domain of the spike protein (S1), and a lateral flow immunoassay (LFI) based on full-length spike protein. Neutralization assays with live SARS-CoV-2 were performed. One thousand four hundred and seventy-seven individuals were included comprising 112 SARS-CoV-2 positives (defined as a positive real-time PCR result; prevalence 7.6%). IgG seroconversion occurred between day 0 and day 21. While the ELISAs showed sensitivities of 88.4% for RBD, 89.3% for S1, and 72.9% for N protein, the specificity was above 94% for all tests. Out of 54 SARS-CoV-2 positive individuals, 96.3% showed full neutralization of live SARS-CoV-2 at serum dilutions ≥ 1:16, while none of the 6 SARS-CoV-2-negative sera revealed neutralizing activity. ELISAs targeting RBD and S1 protein of SARS-CoV-2 are promising immunoassays which shall be further evaluated in studies verifying diagnostic accuracy and protective immunity against SARS-CoV-2.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA, ready to use solution
Sigma-Aldrich
Anti-Human IgM (μ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody