Merck
Co-regulation and function of FOXM1/RHNO1 bidirectional genes in cancer.
eLife (2021-04-24)
Carter J Barger, Linda Chee, Mustafa Albahrani, Catalina Munoz-Trujillo, Lidia Boghean, Connor Branick, Kunle Odunsi, Ronny Drapkin, Lee Zou, Adam R Karpf
ABSTRAKT
The FOXM1 transcription factor is an oncoprotein and a top biomarker of poor prognosis in human cancer. Overexpression and activation of FOXM1 is frequent in high-grade serous carcinoma (HGSC), the most common and lethal form of human ovarian cancer, and is linked to copy number gains at chromosome 12p13.33. We show that FOXM1 is co-amplified and co-expressed with RHNO1, a gene involved in the ATR-Chk1 signaling pathway that functions in the DNA replication stress response. We demonstrate that FOXM1 and RHNO1 are head-to-head (i.e., bidirectional) genes (BDG) regulated by a bidirectional promoter (BDP) (named F/R-BDP). FOXM1 and RHNO1 each promote oncogenic phenotypes in HGSC cells, including clonogenic growth, DNA homologous recombination repair, and poly-ADP ribosylase inhibitor resistance. FOXM1 and RHNO1 are one of the first examples of oncogenic BDG, and therapeutic targeting of FOXM1/RHNO1 BDG is a potential therapeutic approach for ovarian and other cancers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Carboplatin