Merck
  • Immunoglobulin G antibody response to the Sputnik V vaccine: previous SARS-CoV-2 seropositive individuals may need just one vaccine dose.
Immunoglobulin G antibody response to the Sputnik V vaccine: previous SARS-CoV-2 seropositive individuals may need just one vaccine dose.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2021-08-04)
Franklin Claro, Douglas Silva, Melissa Rodriguez, Hector Rafael Rangel, Jacobus H de Waard
ABSTRAKT
We evaluated the immunoglobulin (Ig) G antibody response against the nucleocapsid protein (NP) and the receptor-binding domain (RBD) of the spike protein of SARS-CoV-2 in a cohort of 86 individuals in Venezuela, before and after receiving the Sputnik V vaccine. Antibody responses against NP and RBD were determined with an enzyme-linked immunosorbent assay just before, 3 weeks after the first, and 6 weeks after the second dose of the vaccine. Before vaccination, 59 individuals were seronegative, and 27 seropositive for NP and/or RBD. Of the seronegative cohort, 42% did not develop an IgG immune response against RBD after the first vaccine dose, but 100% had a strong IgG response after 2 doses. All seropositive individuals developed a strong IgG antibody response against RBD after the first vaccine dose, with antibody levels ∼40% higher than seronegative individuals who had received 2 doses. Previously seropositive subjects showed no significant increase in IgG antibody response against RBD after the second vaccine dose. We demonstrate that 2 doses of the Sputnik V vaccine triggered antibody response in all study individuals. The second Sputnik V dose had no impact on IgG response for those seropositive for SARS-CoV-2 antigens before vaccination.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate