Merck
  • Author Correction: The epicardium obscures interpretations on endothelial-to-mesenchymal transition in the mouse atrioventricular canal explant assay.
Author Correction: The epicardium obscures interpretations on endothelial-to-mesenchymal transition in the mouse atrioventricular canal explant assay.
Scientific reports (2020-04-02)
Nathan Criem, An Zwijsen
ABSTRAKT
An amendment to this paper has been published and can be accessed via a link at the top of the paper.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Kolagen typu I, ogon szczura, 100 mg, solution, BioReagent, suitable for cell culture