Merck
Ezh1 regulates expression of Cpg15/Neuritin in mouse cortical neurons.
Drug discoveries & therapeutics (2021-03-09)
Shun Utsunomiya, Yusuke Kishi, Masafumi Tsuboi, Daichi Kawaguchi, Yukiko Gotoh, Manabu Abe, Kenji Sakimura, Kazuma Maeda, Hiroshi Takemoto
ABSTRAKT
Immature neurons undergo morphological and physiological maturation in order to establish neuronal networks. During neuronal maturation, a large number of genes change their transcriptional levels, and these changes may be mediated by chromatin modifiers. In this study, we found that the level of Ezh1, a component of Polycomb repressive complex 2 (PRC2), increases during neuronal maturation in mouse neocortical culture. In addition, conditional knockout of Ezh1 in post-mitotic excitatory neurons leads to downregulation of a set of genes related to neuronal maturation. Moreover, the locus encoding Cpg15/Neuritin (Nrn1), which is regulated by neuronal activity and implicated in stabilization and maturation of excitatory synapses, is a direct target of Ezh1 in cortical neurons. Together, these results suggest that elevated expression of Ezh1 contributes to maturation of cortical neurons.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw trimetylo-histonowi H3 (Lys27), Upstate®, from rabbit