- Targeting the spliceosome through RBM39 degradation results in exceptional responses in high-risk neuroblastoma models.
Targeting the spliceosome through RBM39 degradation results in exceptional responses in high-risk neuroblastoma models.
Science advances (2021-11-18)
Shivendra Singh, Waise Quarni, Maria Goralski, Shibiao Wan, Hongjian Jin, Lee-Ann Van de Velde, Jie Fang, Qiong Wu, Ahmed Abu-Zaid, Tingting Wang, Ravi Singh, David Craft, Yiping Fan, Thomas Confer, Melissa Johnson, Walter J Akers, Ruoning Wang, Peter J Murray, Paul G Thomas, Deepak Nijhawan, Andrew M Davidoff, Jun Yang
PMID34788094
ABSTRAKT
[Figure: see text].
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
LookOut® Mycoplasma PCR Detection Kit, Optimized for use with JumpStart™ Taq DNA Polymerase, D9307.