Merck
  • Crosstalk Between RPE Cells and Choroidal Endothelial Cells via the ANXA1/FPR2/SHP2/NLRP3 Inflammasome/Pyroptosis Axis Promotes Choroidal Neovascularization.
Crosstalk Between RPE Cells and Choroidal Endothelial Cells via the ANXA1/FPR2/SHP2/NLRP3 Inflammasome/Pyroptosis Axis Promotes Choroidal Neovascularization.
Inflammation (2021-10-02)
Manhui Zhu, Ying Wang, Linling Zhu, Shu Du, Zhenzhen Wang, Yuting Zhang, Yang Guo, Yuanyuan Tu, E Song
ABSTRAKT
One type of age-related macular degeneration (AMD), neovascular (nAMD), characterized by choroidal neovascularization (CNV), accounts for the majority of the severe central vision impairment associated with AMD. Endothelial cells (ECs) in direct contact with retinal pigment epithelial (RPE) cells are more prone to the pathological angiogenesis involved in CNV. Herein, we investigated the effect of crosstalk between RPE cells and choroidal endothelial cells (CECs) via the ANXA1/FPR2/NLRP3 inflammasome/pyroptosis axis on the development of choroidal neovascularization (CNV) in vitro and in vivo. ANXA1 expression and secretion from ARPE-19 cells were upregulated by hypoxia. FPR2 expression, especially on the plasma membrane, in HCECs was upregulated under hypoxic conditions. ANXA1 secreted from ARPE-19 cells inhibited NLRP3 inflammasome activation and NLRP3 inflammasome-mediated pyroptosis in HCECs by activating the FPR2/SHP2 axis. Moreover, ANXA1 secreted by ARPE-19 cells promoted behaviors of HCECs, including proliferation, migration, and tube formation, by activating the FPR2/SHP2 axis and inhibiting NLRP3 inflammasome-mediated pyroptosis. Inhibiting the upregulated ANXA1/FPR2/SHP2/NLRP3 inflammasome/pyroptosis axis decreased the volume of CNV. Our data suggest that the crosstalk between RPE cells and CECs via the ANXA1/FPR2/NLRP3 inflammasome/pyroptosis axis promotes CNV. This finding could identify a potential target for the prevention and treatment of CNV.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
RIPA Buffer
Roche
7-Deaza-2′-deoksy-5′-trifosforan guanozyny, 95% (7-Deaza-dGTP, HPLC), ≤4% (7-Deaza-dGDP, HPLC), solution, 10 mM
Roche
Zestaw do wykrywania śmierci komórek in situ, fluoresceina, sufficient for ≤50 tests, suitable for detection