Merck
Sepsis induces muscle atrophy by inhibiting proliferation and promoting apoptosis via PLK1-AKT signalling.
Journal of cellular and molecular medicine (2021-09-14)
Ying-Ya Cao, Zhen Wang, Tao Yu, Yuan Zhang, Zhong-Han Wang, Zi-Meng Lu, Wei-Hua Lu, Jian-Bo Yu
ABSTRAKT
Sepsis and sepsis-induced skeletal muscle atrophy are common in patients in intensive care units with high mortality, while the mechanisms are controversial and complicated. In the present study, the atrophy of skeletal muscle was evaluated in sepsis mouse model as well as the apoptosis of muscle fibres. Sepsis induced atrophy of skeletal muscle and apoptosis of myofibres in vivo and in vitro. In cell-based in vitro experiments, lipopolysaccharide (LPS) stimulation also inhibited the proliferation of myoblasts. At the molecular level, the expression of polo-like kinase 1 (PLK1) and phosphorylated protein kinase B (p-AKT) was decreased. Overexpression of PLK1 partly rescued LPS-induced apoptosis, proliferation suppression and atrophy in C2C12 cells. Furthermore, inhibiting the AKT pathway deteriorated LPS-induced atrophy in PLK1-overexpressing C2C12 myotubes. PLK1 was found to participate in regulating apoptosis and E3 ubiquitin ligase activity in C2C12 cells. Taken together, these results indicate that sepsis induces skeletal muscle atrophy by promoting apoptosis of muscle fibres and inhibiting proliferation of myoblasts via regulation of the PLK1-AKT pathway. These findings enhance understanding of the mechanism of sepsis-induced skeletal muscle atrophy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-PLK1, klon 35-206, clone 35-206, Upstate®, from mouse