Merck
Dual-sided Voltage-sensitive Dye Imaging of Leech Ganglia.
Bio-protocol (2018-03-30)
Yusuke Tomina, Daniel A Wagenaar
ABSTRAKT
In this protocol, we introduce an effective method for voltage-sensitive dye (VSD) loading and imaging of leech ganglia as used in Tomina and Wagenaar (2017). Dissection and dye loading procedures are the most critical steps toward successful whole-ganglion VSD imaging. The former entails the removal of the sheath that covers neurons in the segmental ganglion of the leech, which is required for successful dye loading. The latter entails gently flowing a new generation VSD, VF2.1(OMe).H, onto both sides of the ganglion simultaneously using a pair of peristaltic pumps. We expect the described techniques to translate broadly to wide-field VSD imaging in other thin and relatively transparent nervous systems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)