Merck
Use of drugs to study role of microtubule assembly dynamics in living cells.
Methods in enzymology (1998-09-30)
M A Jordan, L Wilson
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Vinblastine sulfate salt, 96-102% (HPLC)