Merck
NogoA-expressing astrocytes limit peripheral macrophage infiltration after ischemic brain injury in primates.
Nature communications (2021-11-27)
Anthony G Boghdadi, Joshua Spurrier, Leon Teo, Mingfeng Li, Mario Skarica, Benjamin Cao, William C Kwan, Tobias D Merson, Susan K Nilsson, Nenad Sestan, Stephen M Strittmatter, James A Bourne
ABSTRAKT
Astrocytes play critical roles after brain injury, but their precise function is poorly defined. Utilizing single-nuclei transcriptomics to characterize astrocytes after ischemic stroke in the visual cortex of the marmoset monkey, we observed nearly complete segregation between stroke and control astrocyte clusters. Screening for the top 30 differentially expressed genes that might limit stroke recovery, we discovered that a majority of astrocytes expressed RTN4A/ NogoA, a neurite-outgrowth inhibitory protein previously only associated with oligodendrocytes. NogoA upregulation on reactive astrocytes post-stroke was significant in both the marmoset and human brain, whereas only a marginal change was observed in mice. We determined that NogoA mediated an anti-inflammatory response which likely contributes to limiting the infiltration of peripheral macrophages into the surviving parenchyma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Roche
cOmplete, Mini, Koktajl inhibitorów proteaz bez EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Roche
cOmplete, Mini koktajl inhibitorów proteaz, Tablets provided in a glass vial
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M