Merck
Dynamic Behavior in Enzyme-Polymer Surfactant Hydrogel Films.
Advanced materials (Deerfield Beach, Fla.) (2015-12-18)
Kamendra P Sharma, Robert Harniman, Thomas Farrugia, Wuge H Briscoe, Adam W Perriman, Stephen Mann
ABSTRAKT
Dynamic protein-polymer surfactant films are highly hydrophilic and show a soft solid to hydrogel transition upon hydration to produce a swollen hydrogel. An unusual reversible autospreading/self-folding response is observed when the water-saturated films are transferred from water into air.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution