Merck
  • Insight into the Characteristics of Novel Desmin-Immunopositive Perivascular Cells of the Anterior Pituitary Gland Using Transmission and Focused Ion Beam Scanning Electron Microscopy.
Insight into the Characteristics of Novel Desmin-Immunopositive Perivascular Cells of the Anterior Pituitary Gland Using Transmission and Focused Ion Beam Scanning Electron Microscopy.
International journal of molecular sciences (2021-08-28)
Depicha Jindatip, Rebecca Wan-Yan Poh, Ken Fujiwara
ABSTRAKT
Recently, another new cell type was found in the perivascular space called a novel desmin-immunopositive perivascular (DIP) cell. However, the differences between this novel cell type and other nonhormone-producing cells have not been clarified. Therefore, we introduced several microscopic techniques to gain insight into the morphological characteristics of this novel DIP cell. We succeeded in identifying novel DIP cells under light microscopy using desmin immunocryosection, combining resin embedding blocks and immunoelectron microscopy. In conventional transmission electron microscopy, folliculostellate cells, capsular fibroblasts, macrophages, and pericytes presented a flat cisternae of rough endoplasmic reticulum, whereas those of novel DIP cells had a dilated pattern. The number of novel DIP cells was greatest in the intact rats, though nearly disappeared under prolactinoma conditions. Additionally, focused ion beam scanning electron microscopy showed that these novel DIP cells had multidirectional processes and some processes reached the capillary, but these processes did not tightly wrap the vessel, as is the case with pericytes. Interestingly, we found that the rough endoplasmic reticulum was globular and dispersed throughout the cytoplasmic processes after three-dimensional reconstruction. This study clearly confirms that novel DIP cells are a new cell type in the rat anterior pituitary gland, with unique characteristics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ≥96%