Merck
  • Native and fluorescent dye-dependent single-DNA molecule microchip dynamics as measured by differential interference contrast microscopy.
Native and fluorescent dye-dependent single-DNA molecule microchip dynamics as measured by differential interference contrast microscopy.
Chemical communications (Cambridge, England) (2011-07-14)
Doori Oh, Seungah Lee, Seong Ho Kang
ABSTRAKT
In a free solution of 10.0 mM Gly-Gly (pH 8.2), the flow directions of native-DNA and DNA molecules intercalated with the fluorescent dye YOYO-1 were reversed in the microchip channel. These results clearly showed that in a confined space and under specific environmental conditions, the fluorescent dye modified the original properties and behavior of the native-DNA molecule.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)