Merck
Synthesis of a 42 residue peptide corresponding to the entire amino acid sequence of human GIP.
Chemical & pharmaceutical bulletin (1985-08-01)
H Yajima, N Fujii, K Akaji, M Sakurai, M Nomizu, K Mizuta, M Aono, M Moriga, K Inoue, R Hosotani
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gastric Inhibitory Polypeptide human, ≥95% (HPLC)