Merck
A father's IgE-mediated contact urticaria from mother's milk.
The Journal of allergy and clinical immunology (2004-02-10)
Soili Mäkinen-Kiljunen, Mikko Plosila
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Casein sodium salt from bovine milk