Merck
Altered cytoskeletal status in the transition from proneural to mesenchymal glioblastoma subtypes.
Scientific reports (2022-06-15)
Maureen Keller, Magdalena Blom, Lei Liu Conze, Min Guo, Daniel Hägerstrand, Pontus Aspenström
ABSTRAKT
Glioblastoma is a highly aggressive brain tumor with poor patient prognosis. Treatment outcomes remain limited, partly due to intratumoral heterogeneity and the invasive nature of the tumors. Glioblastoma cells invade and spread into the surrounding brain tissue, and even between hemispheres, thus hampering complete surgical resection. This invasive motility can arise through altered properties of the cytoskeleton. We hypothesize that cytoskeletal organization and dynamics can provide important clues to the different malignant states of glioblastoma. In this study, we investigated cytoskeletal organization in glioblastoma cells with different subtype expression profiles, and cytoskeletal dynamics upon subtype transitions. Analysis of the morphological, migratory, and invasive properties of glioblastoma cells identified cytoskeletal components as phenotypic markers that can serve as diagnostic or prognostic tools. We also show that the cytoskeletal function and malignant properties of glioblastoma cells shift during subtype transitions induced by altered expression of the neurodevelopmental transcription factor SOX2. The potential of SOX2 re-expression to reverse the mesenchymal subtype into a more proneural subtype might open up strategies for novel glioblastoma treatments.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid